accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNH2OH3C

CAS RN 2990-02-5

IUPAC Name: (2,4-dimethylphenyl)urea
Synonyms: 2,4-dimethylphenylurea
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: GKEHHLJOYZDXMI-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(N)=O)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results