accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 29943-42-8

IUPAC Name: oxan-4-one
Synonyms: 4H-pyran-4-one, tetrahydro-
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: JMJRYTGVHCAYCT-UHFFFAOYSA-N
SMILES: O=C1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)