accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 29968-78-3

NH3NOO

CAS RN 29968-78-3

IUPAC Name: 2-(4-nitrophenyl)ethan-1-aminium
Synonyms: 2-(4-nitrophenyl)ethanaminium
Molecular Weight (g/mol): 167.19
Molecular Formula: C8H11N2O2
InChi Key: IOXOZOPLBFXYLM-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)