accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 300-76-5

H3COPOOH3COBrClClBr

CAS RN 300-76-5

IUPAC Name: 1,2-dibromo-2,2-dichloroethyl dimethyl phosphate
Synonyms: naled bromchlophos caswell no. 586 epa pesticide chemical code 034401 fosbrom ortho-dibrom bromex (insecticide) BRP hibrom dibromfos
Molecular Weight (g/mol): 380.78
Molecular Formula: C4H7Br2Cl2O4P
InChi Key: BUYMVQAILCEWRR-UHFFFAOYNA-N
SMILES: COP(=O)(OC)OC(Br)C(Cl)(Cl)Br
Molecular Weight (g/mol) 
  • (3)
  • (6)
Solvent or Matrix 
  • (4)
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)