accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 300-87-8

CH3ONH3C

CAS RN 300-87-8

IUPAC Name: 3,5-dimethyl-1,2-oxazole
Synonyms: isoxazole, 3,5-dimethyl-
Molecular Weight (g/mol): 97.12
Molecular Formula: C5H7NO
InChi Key: FICAQKBMCKEFDI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results