accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NN

CAS RN 3001-72-7

IUPAC Name: 2H,3H,4H,6H,7H,8H-pyrrolo[1,2-a]pyrimidine
Synonyms: DBN
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: SGUVLZREKBPKCE-UHFFFAOYSA-N
SMILES: C1CN2CCCN=C2C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)