accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NN

CAS RN 3002-81-1

IUPAC Name: 5,6-dimethyl-1,10-phenanthroline
Synonyms: 5,6-dimethyl-1,10-phenanthroline
Molecular Weight (g/mol): 208.26
Molecular Formula: C14H12N2
InChi Key: BRPQDJPJBCQFSR-UHFFFAOYSA-N
SMILES: CC1=C(C)C2=CC=CN=C2C2=NC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results