accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOF

CAS RN 30058-40-3

IUPAC Name: 2-fluorobenzene-1-sulfonamide
Synonyms: 2-fluorobenzenesulfonamide
Molecular Weight (g/mol): 175.18
Molecular Formula: C6H6FNO2S
InChi Key: WFLBWYLZCQOPCA-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results