accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNNCl

CAS RN 300816-22-2

IUPAC Name: 12-chloro-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraene
Synonyms: 12-chloro-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraene
Molecular Weight (g/mol): 210.68
Molecular Formula: C9H7ClN2S
InChi Key: JKXJMJPJOYMQSK-UHFFFAOYSA-N
SMILES: ClC1=C2C3=C(CCC3)SC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results