accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 301-00-8

IUPAC Name: methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Synonyms: methyl linolenate methyl octadecatrienoate
Molecular Weight (g/mol): 292.46
Molecular Formula: C19H32O2
InChi Key: DVWSXZIHSUZZKJ-YSTUJMKBSA-N
SMILES: CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results