accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PbH3CCH3OOH3CCH3OO

CAS RN 301-08-6

IUPAC Name: λ²-lead(2+) bis(2-ethylhexanoate)
Synonyms: λ²-lead(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 493.6
Molecular Formula: C16H30O4Pb
InChi Key: RUCPTXWJYHGABR-UHFFFAOYNA-L
SMILES: [Pb++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results