accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 301-10-0

IUPAC Name: λ²-tin(2+) bis(2-ethylhexanoate)
Synonyms: tin(II) 2-ethylhexanoate
Molecular Weight (g/mol): 405.12
Molecular Formula: C16H30O4Sn
InChi Key: KSBAEPSJVUENNK-UHFFFAOYNA-L
SMILES: [Sn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)