accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3CNH3CCH3CH3

CAS RN 3026-69-5

IUPAC Name: dimethyldioctylazanium bromide
Synonyms: dimethyldioctylazanium bromide
Molecular Weight (g/mol): 350.43
Molecular Formula: C18H40BrN
InChi Key: APTVNWGLSRAOFJ-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[N+](C)(C)CCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results