accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30273-00-8

IUPAC Name: 2,4-dichloro-6-methylaniline
Synonyms: 2,4-dichloro-6-methylaniline
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: UAISVUGQLKXPFF-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(Cl)=C1N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results