accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3OH3COH3CH3CH3CH3CH3CH3CH3CH3CH3CCH3H3C(E)(E)(E)(E)(E)(E)(E)(E)(E)

CAS RN 303-98-0

IUPAC Name: 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Synonyms: coenzyme-Q10 inokiten all-trans-ubiquinone coenzyme Q(sub 10) unbiquinone unispheres Q 10 ubidecarenone ubidecarenona luvacor ubiquinone Q10
Molecular Weight (g/mol): 863.37
Molecular Formula: C59H90O4
InChi Key: ACTIUHUUMQJHFO-UPTCCGCDSA-N
SMILES: COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)