accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 303144-44-7

OONNN

CAS RN 303144-44-7

IUPAC Name: 1-(pyrimidin-2-yl)piperidine-4-carboxylate
Synonyms: 1-(pyrimidin-2-yl)piperidine-4-carboxylate
Molecular Weight (g/mol): 206.23
Molecular Formula: C10H12N3O2
InChi Key: DYVXURZASBPYFR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)