accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOOHH2NCl

CAS RN 3032-32-4

IUPAC Name: 2-amino-3,6-dichlorobenzoic acid
Synonyms: 2-amino-3,6-dichlorobenzoic acid
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: MMIREQFMYZQWLU-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC(Cl)=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results