accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3034-55-7

IUPAC Name: 5-bromo-1,3-thiazole
Synonyms: 5-bromo-1,3-thiazole
Molecular Weight (g/mol): 164.02
Molecular Formula: C3H2BrNS
InChi Key: DWUPYMSVAPQXMS-UHFFFAOYSA-N
SMILES: BrC1=CN=CS1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results