accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH

CAS RN 3034-86-4

IUPAC Name: methyl 4-ethynylbenzoate
Synonyms: methyl 4-ethynylbenzoate
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: JPGRSTBIEYGVNO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results