accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFN

CAS RN 3038-47-9

IUPAC Name: 2-[2-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: QXDCZSJGEUSERL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1CC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results