accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 304-20-1

IUPAC Name: hydrogen 1-hydrazinylphthalazine chloride
Synonyms: hydralazine
Molecular Weight (g/mol): 196.64
Molecular Formula: C8H9ClN4
InChi Key: ZUXNZUWOTSUBMN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=C2C=CC=CC2=CN=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results