CAS RN 3040-56-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Di-Beta,Beta'-Chloroethylphosphoric Acid, TRC
CAS: 3040-56-0 Molecular Formula: C4H9Cl2O4P Molecular Weight (g/mol): 222.99 Synonym: Ethanol, 2-chloro-, hydrogen phosphate (7CI, 8CI, 9CI),Bis(2-chloroethyl) hydrogen phosphate,Bis(2-chloroethyl) phosphate,Bis(chloroethyl) phosphate,Bis(β-chloroethyl) phosphate,Bis(β-chloroethyl) phosphoric acid,Di-2-Chloroethyl phosphate,Di-β,β'-Chloroethylphosphoric acid IUPAC Name: bis(2-chloroethyl) hydrogen phosphate SMILES: OP(=O)(OCCCl)OCCCl