accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOO

CAS RN 3041-16-5

IUPAC Name: 1,4-dioxan-2-one
Synonyms: 1,4-dioxan-2-one
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: VPVXHAANQNHFSF-UHFFFAOYSA-N
SMILES: O=C1COCCO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)