accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30414-54-1

H3COOOCH3

CAS RN 30414-54-1

IUPAC Name: methyl 3-oxohexanoate
Synonyms: methyl 3-oxohexanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: SJPCQNABHNCLPB-UHFFFAOYSA-N
SMILES: CCCC(=O)CC(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)