accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFFOOFFFOOFFFOY

CAS RN 304851-95-4

IUPAC Name: tris(trifluoroacetic acid) hydrate yttrium
Synonyms: tris(trifluoroacetic acid) hydrate yttrium
Molecular Weight (g/mol): 448.99
Molecular Formula: C6H5F9O7Y
InChi Key: RTYFWTAMUWPTNO-UHFFFAOYSA-N
SMILES: O.[Y].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results