accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 304884-73-9

IUPAC Name: N-(2,6-dichlorophenyl)-2-fluorobenzamide
Synonyms: N-(2,6-dichlorophenyl)-2-fluorobenzamide
Molecular Weight (g/mol): 284.11
Molecular Formula: C13H8Cl2FNO
InChi Key: CSQKIRJRBCAPIY-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)NC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results