accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SBr

CAS RN 30506-30-0

IUPAC Name: 1-bromo-4-(ethylsulfanyl)benzene
Synonyms: 1-bromo-4-(ethylsulfanyl)benzene
Molecular Weight (g/mol): 217.12
Molecular Formula: C8H9BrS
InChi Key: CTFHVAKXUMMGOC-UHFFFAOYSA-N
SMILES: CCSC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results