accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2O

CAS RN 30525-89-4

IUPAC Name: formaldehyde
Synonyms: formaldehyde caswell no. 633 caswell no. 465 formic aldehyde formaldehyde, gas formol) formaldehyde (formalin superlysoform paraformaldehydum methylene oxide BFV
Molecular Weight (g/mol): 30.03
Molecular Formula: CH2O
InChi Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES: C=O
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (18)
  • (4)
Physical Form 
  • (5)
  • (3)
  • (6)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (5)
  • (3)
Grade 
  • (5)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (7)
  • (7)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (18)
  • (4)
  • (5)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)