accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30536-19-7

NH2ClNSN

CAS RN 30536-19-7

IUPAC Name: 5-chloro-2,1,3-benzothiadiazol-4-amine
Synonyms: 5-chloro-2,1,3-benzothiadiazol-4-amine
Molecular Weight (g/mol): 185.63
Molecular Formula: C6H4ClN3S
InChi Key: MURNIACGGUSMAP-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC2=NSN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results