accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHN(R)O(S)OOCH3HO

CAS RN 3056-17-5

IUPAC Name: 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: stavudine 3'-deoxy-2'-thymidinene stavudine [usan:ban:inn] STV estavudina [inn-spanish] sanilvudine estavudina stavudinum
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: XNKLLVCARDGLGL-JGVFFNPUSA-N
SMILES: CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)