accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3057-74-7

IUPAC Name: 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Synonyms: 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: MXWMFBYWXMXRPD-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results