accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306-08-1

IUPAC Name: 2-(4-hydroxy-3-methoxyphenyl)acetic acid
Synonyms: homovanillic acid vanilacetic acid homovanilate 4 hydroxy 3 methoxyphenylacetic acid HVA acid, homovanillic 3 methoxy 4 hydroxyphenylacetic acid vanillacetic acid vanilacetate hva - homovanillic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: QRMZSPFSDQBLIX-UHFFFAOYSA-N
SMILES: COC1=CC(CC(O)=O)=CC=C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results