accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306-91-2

IUPAC Name: tetracosafluoro-tetradecahydrophenanthrene
Synonyms: perfluorophenanthrene
Molecular Weight (g/mol): 624.12
Molecular Formula: C14F24
InChi Key: QKENRHXGDUPTEM-UHFFFAOYNA-N
SMILES: FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(C1(F)F)C(F)(F)C(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C21F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results