accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30683-23-9

IUPAC Name: 3-bromopyridine-2-carboxylic acid
Synonyms: 3-bromopyridine-2-carboxylic acid
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: KBDIRPOTVAODSA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results