accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSiCH3NHNH2OCH3

CAS RN 3069-29-2

IUPAC Name: (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine
Synonyms: (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine
Molecular Weight (g/mol): 206.36
Molecular Formula: C8H22N2O2Si
InChi Key: MQWFLKHKWJMCEN-UHFFFAOYSA-N
SMILES: CO[Si](C)(CCCNCCN)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results