accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306934-71-4

NHNNNS

CAS RN 306934-71-4

IUPAC Name: 1-(3-phenyl-1,2,4-thiadiazol-5-yl)-1,4-diazepane
Synonyms: 1-(3-phenyl-1,2,4-thiadiazol-5-yl)-1,4-diazepane
Molecular Weight (g/mol): 260.36
Molecular Formula: C13H16N4S
InChi Key: IENGAZSTLAONAR-UHFFFAOYSA-N
SMILES: C1CNCCN(C1)C1=NC(=NS1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results