accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306934-89-4

IUPAC Name: (6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl)methanol
Synonyms: (6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl)methanol
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: YLADDVZIEBXWQU-UHFFFAOYSA-N
SMILES: OCC1=C2OCOCC2=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results