accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306935-05-7

IUPAC Name: 1-[5-methyl-2-(trifluoromethyl)furan-3-yl]methanamine
Synonyms: 1-[5-methyl-2-(trifluoromethyl)furan-3-yl]methanamine
Molecular Weight (g/mol): 179.14
Molecular Formula: C7H8F3NO
InChi Key: ROYYYTOVBUUPDX-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=C(O1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results