accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306935-14-8

IUPAC Name: 1-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine
Synonyms: 1-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine
Molecular Weight (g/mol): 246.33
Molecular Formula: C12H14N4S
InChi Key: UMFMHSLRNJBGKO-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=NC(=NS1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results