accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306935-39-7

IUPAC Name: 1-(2H-1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
Synonyms: 1-(2H-1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
Molecular Weight (g/mol): 260.17
Molecular Formula: C11H7F3O4
InChi Key: FDLJUCOQPAGQNY-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results