accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306936-14-1

IUPAC Name: 2,5-dimethyl-1-[(thiophen-2-yl)methyl]-1H-pyrrole-3-carboxylic acid
Synonyms: 2,5-dimethyl-1-(thiophen-2-ylmethyl)pyrrole-3-carboxylic acid
Molecular Weight (g/mol): 235.3
Molecular Formula: C12H13NO2S
InChi Key: MYRBXKXOHIDTFU-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(C)N1CC1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results