accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 306936-15-2

IUPAC Name: 2,5-dimethyl-1-[(pyridin-4-yl)methyl]-1H-pyrrole-3-carboxylic acid
Synonyms: 2,5-dimethyl-1-(pyridin-4-ylmethyl)pyrrole-3-carboxylic acid
Molecular Weight (g/mol): 230.27
Molecular Formula: C13H14N2O2
InChi Key: IHQUENCAKSKBOG-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(C)N1CC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results