CAS RN 307-24-4
IUPAC Name:
undecafluorohexanoic acid
Synonyms:
undecafluorohexanoic acid
Molecular Weight (g/mol):
314.05
Molecular Formula:
C6HF11O2
InChi Key:
PXUULQAPEKKVAH-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
2
of
2
results
1
Perfluorohexanoic Acid (>90%), TRC
CAS: 307-24-4 Molecular Formula: C6 H F11 O2 Molecular Weight (g/mol): 314.05 Synonym: Hexanoic acid, undecafluoro- (6CI,7CI,8CI,9CI),2,2,3,3,4,4,5,5,6,6,6-Undecafluorohexanoic acid,NSC 5213,PFHxA,Perfluoro-1-pentanecarboxylic acid,Perfluorohexanoic acid IUPAC Name: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
2
Perfluorohexanoic Acid (50μg/mL in Methanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.