accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 307-55-1

IUPAC Name: tricosafluorododecanoic acid
Synonyms: perfluorododecanoic acid
Molecular Weight (g/mol): 614.1
Molecular Formula: C12HF23O2
InChi Key: CXGONMQFMIYUJR-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results