accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2BrZnBr

CAS RN 307496-31-7

IUPAC Name: (3-bromophenyl)methanide; bromozincylium
Synonyms: (3-bromophenyl)methanide; bromozincylium
Molecular Weight (g/mol): 315.31
Molecular Formula: C7H6Br2Zn
InChi Key: FBSUMSXZNCNBLE-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results