accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CH3CCH3H3CBOOCH3H3CCH3CH3CH3(E/Z)

CAS RN 307531-74-4

IUPAC Name: 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hept-1-en-1-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hept-1-en-1-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 350.11
Molecular Formula: C19H36B2O4
InChi Key: KBMWLQPDFRGVTD-UHFFFAOYSA-N
SMILES: CCCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results