accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2SFOO

CAS RN 30827-99-7

IUPAC Name: hydrogen 4-(2-aminoethyl)benzene-1-sulfonyl fluoride chloride
Synonyms: hydrogen aebsf chloride
Molecular Weight (g/mol): 239.69
Molecular Formula: C8H11ClFNO2S
InChi Key: WRDABNWSWOHGMS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=C(C=C1)S(F)(=O)=O
Molecular Weight (g/mol) 
  • (13)
  • (2)
  • (2)
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
Physical Form 
  • (3)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (13)
  • (2)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (8)