accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3SCH3H3C

CAS RN 3084-53-5

IUPAC Name: trimethylsulfanium bromide
Synonyms: trimethylsulfonium bromide
Molecular Weight (g/mol): 157.07
Molecular Formula: C3H9BrS
InChi Key: GOTIICCWNAPLMN-UHFFFAOYSA-M
SMILES: [Br-].C[S+](C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results