accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3088-42-4

HOONHN

CAS RN 3088-42-4

IUPAC Name: 2-[(2-cyanoethyl)amino]acetic acid
Synonyms: glycine, N-(2-cyanoethyl)-
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: KZUBZCHAWPDYQX-UHFFFAOYSA-N
SMILES: OC(=O)CNCCC#N
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)