accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CCH3MgBr

CAS RN 30897-86-0

IUPAC Name: magnesium(2+) 2,5-dimethylbenzen-1-ide bromide
Synonyms: magnesium(2+) 2,5-dimethylbenzen-1-ide bromide
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: OOJAWTIYZUSKLZ-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=CC=C(C)[C-]=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results